missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Ponceau 4R

Product Code. 16412419
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16412419 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16412419 Supplier MedChemExpress Supplier No. HYD0193
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ponceau 4R is a synthetic colourant that may be used as a food colouring.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Product Type Dye Reagents
Quantity 1 g
Shipping Condition Room temperature
Synonym Acid Red 18 ∣ New Coccine
CAS 2611-82-7
Molecular Weight (g/mol) 604.47 g/mol
Solubility DMSO : 100 mg/mL (165.43 mM; Need ultrasonic) ∣ H2O : 25 mg/mL (41.36 mM; Need ultrasonic)
Target HBV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.