missing translation for 'onlineSavingsMsg'
Learn More

Ponceau S, 85%, MP Biomedicals Inc

Product Code. 11486061 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g

Product Code. 11486061

Brand: MP Biomedicals 0219064405

Please to purchase this item. Need a web account? Register with us today!

Chemical Identifiers

CAS 6226-79-5
Molecular Formula C22H12N4Na4O13S4
Molecular Weight (g/mol) 760.552
InChI Key VSXKEUCERCWGKF-STNZDNLRSA-J
Synonym 3-Hydroxy-4-(2-sulfo-4-(4-sulfophenylazo)phenylazo)-2, 7-naphthalene-disulfonic acid tetrasodium salt, C.I. 27195
PubChem CID 11320219
IUPAC Name tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate
SMILES C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Assay Percent Range ∽ 85%
Formula Weight 760.6
Quantity 5 g
Chemical Name or Material Ponceau S, Protein stain for electrophoresis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.