missing translation for 'onlineSavingsMsg'
Learn More

Ponceau S, Electrophoresis Grade

Product Code. 15483449
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483449 10 g 10g
15493449 50 g 50g
15403459 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15483449 Supplier Thermo Scientific Alfa Aesar Supplier No. J60744.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protein stain for electrophoresis | CAS: 6226-79-5 | C22H12N4Na4O13S4 | 760.552 g/mol

Ponceau S dye is used to make a stain for rapid reversible staining of protein bands on nitrocellulose or PVDF membranes (Western blots) and also for staining protein on cellulose acetate membranes. It is also suitable for electrophoresis, azo dye for reversible protein staining. This stain can also be used as a fiducial marker in mass spectrometric imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ponceau S dye is used to make a stain for rapid reversible staining of protein bands on nitrocellulose or PVDF membranes (Western blots) and also for staining protein on cellulose acetate membranes. It is also suitable for electrophoresis, azo dye for reversible protein staining. This stain can also be used as a fiducial marker in mass spectrometric imaging.

Solubility
Soluble in water (10 g/L) at 20°C. Insoluble in ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away fom strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6226-79-5
Molecular Formula C22H12N4Na4O13S4
Molecular Weight (g/mol) 760.552
MDL Number MFCD00003892
InChI Key VSXKEUCERCWGKF-STNZDNLRSA-J
Synonym C.I. 27195
PubChem CID 11320219
IUPAC Name tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate
SMILES C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Color Brown to Red
Solubility Information Soluble in water (10g/L) at 20°C. Insoluble in ether.
Quantity 10 g
Physical Form Powder
Formula Weight 760.58
Grade Electrophoresis
Chemical Name or Material Ponceau S
compliance-icons
Product Identifier
  • Ponceau S, Electrophoresis Grade
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Ponceau S

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.