missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ POP-7™ Polymer For 3500 Dx Series

Product Code. 15978816 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Samples:
384
960 Samples
Unit Size:
384 tests
960 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Samples unitSize
15978816 384 384 tests
15998816 960 Samples 960 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15978816 Supplier Applied Biosystems™ Supplier No. 4393709

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

384 Samples

Conveniently offered in easy to use pouch packages

  • Choose either 384 or 960 samples
TRUSTED_SUSTAINABILITY

Specifications

Description POP-7™ Polymer, 384 Samples, For 3500 Dx Series
For Use With (Equipment) 3500 Dx Series
No. of Samples 384
Product Type POP-7™ Polymer
Sample Volume 384 Sample
Size 1 Kit

For in-vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.