Learn More
DWK Life Sciences™ Porcelain Desiccator Plate
Product Code. 10672062 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric) Outer:
90 mm
140 mm
190 mm
235 mm
280 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
10672062 280 mm Each
10409542 140 mm Each
10325011 190 mm Each
10099820 235 mm Each
10020091 90 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10672062 Supplier DWK Life Sciences™ Supplier No. 297256909
In Stock
Add to basket
Add to basket
The desiccator insert is a sieve plate on which the substance to be dried is placed in an appropriate container.
The desiccator insert is a sieve plate that separates the desiccator base into two areas - there is a desiccant in the lower part and the substance to be dried is placed in the upper part. The substance to be dried is placed on the sieve plate in an appropriate container for drying.
  • The heat-resistant material allows hot samples to be placed into the desiccator
  • Made of heat-resistant material
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Outer 280 mm
Material Porcelain
For Use With (Application) Drying of materials