missing translation for 'onlineSavingsMsg'
Learn More

Porfiromycin, TRC

Product Code. 30133736
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
5 mg
10 mg
missing translation for 'unitSize':
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30133736 5 mg 5mg
30003407 1 mg 1mg
30012799 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30133736 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCP6894505MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 801-52-5
Molecular Formula C16 H20 N4 O5
Molecular Weight (g/mol) 348.35
Synonym Azirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione, 6-amino-8-[[(aminocarbonyl)oxy]methyl]-1,1a,2,8,8a,8b-hexahydro-8a-methoxy-1,5-dimethyl-, (1aS,8S,8aR,8bS)-, Azirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione, 6-amino-1,1a,2,8,8a,8b-hexahydro-8-(hydroxymethyl)-8a-methoxy-1,5-dimethyl-, carbamate (ester) (8CI), Azirino[2',3':3,4]pyrrolo[1,2-a]indole-4,7-dione, 6-amino-8-[[(aminocarbonyl)oxy]methyl]-1,1a,2,8,8a,8b-hexahydro-8a-methoxy-1,5-dimethyl-, [1aS-(1aα,8β,8aα,8bα)]-, ENT 50825, Methyl mitomycin C, Methylmitomycin, N-Methylmitomycin C, NCI 56410, NSC 56410, Porfiromycin, Porfiromycine, Regamycin, U 14743
SMILES CO[C@@]12[C@H](COC(=O)N)C3=C(N1C[C@H]4[C@@H]2N4C)C(=O)C(=C(N)C3=O)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H20N4O5/c1-6-10(17)13(22)9-7(5-25-15(18)23)16(24-3)14-8(19(14)2)4-20(16)11(9)12(6)21/h7-8,14H,4-5,17H2,1-3H3,(H2,18,23)/t7-,8+,14+,16-,19?/m1/s1
Formula Weight 348.1434
Chemical Name or Material Porfiromycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.