missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ Benzyl Ultra Hydrophobic Interaction Chromatography (HIC) Resin

Product Code. 18486818 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mL
50 mL
250 mL
1 L
Unit Size:
1000mL
250mL
25mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18486818 25 mL 25mL
18426758 50 mL 50mL
18745445 250 mL 250mL
18466728 1 L 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18486818 Supplier Thermo Scientific™ Supplier No. A32569

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ POROS™ Benzyl Hydrophobic Interaction Chromatography (HIC) Resin is a bioprocess high-performance resin engineered for improved impurity removal during large-scale downstream purification of biomolecules.

POROS Benzyl Hydrophobic Interaction Chromatography (HIC) Resin is a bioprocess high-performance resin engineered for improved impurity removal during large-scale downstream purification of biomolecules. This unique resin provides high resolution and capacity while maintaining excellent pressure flow characteristics, helping increase productivity.

POROS Benzyl HIC Resin is designed on the well-established cross‐linked poly(styrene-divinylbenzene) POROS base bead with aromatic hydrophobic benzyl ligand, suited for both bind/elute and flow-through purifications.

Features of POROS Benzyl HIC Resin include:

  • Superior resolution capability while maintaining excellent pressure-flow properties, achieved by combining the high-flow characteristics of a POROS base bead with a 50 micron particle size. Higher flow rates help improve productivity.
  • Unique selectivity due to the aromatic hydrophobic ligand, allowing for purification of a wide range of biomolecules, including therapeutic proteins, antibody fragments, antibody drug conjugates, and enzymes
  • High capacity, reducing the process volume significantly for subsequent steps
  • A rigid polymeric bead with covalent surface chemistry for easier handling and packing with superior physical and chemical stability, allowing cleaning with various clean-in-place (CIP) solutions such as sodium hydroxide and enabling a more robust downstream purification process

These features of POROS Benzyl HIC Resin are a major improvement to the performance of traditional HIC resins, setting the standard for hydrophobic interaction chromatography. POROS Benzyl HIC Resin combines superior and consistent performance, exceptional customer support, strong operational excellence, and a strong legacy based on proven technology from a well-established chromatography resin supplier. This resin comes with complete technical and regulatory support. POROS Benzyl HIC Resin is available in various bulk package sizes for large-scale purification, as well as pre-packed columns for initial process development.

Main characteristics

  • Support matrix: cross-linked poly(styrene-divinylbenzene)
  • Average particle size: 50 μm
  • Ligand: benzyl/aromatic
  • pH range: 1–14
  • Ionic strength range: 0 to 5 M, all common salts
  • Mechanical resistance: 100 bar (1450 psi; 10 MPa)
  • Shipping solution: 18% (v/v) ethanol

Specifications

Product Type Benzyl Ultra Resin
Quantity 25 mL
Particle Size 50 μm
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Product Line POROS
Stationary Phase High-Density Benzyl/Aromatic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.