Learn More
Thermo Scientific™ POROS™ Caprylate RoboColumn™ Pre-packed columns
Shop All Thermo Scientific ProductsDescription
These RoboColumns are pre-packed with POROS Caprylate Mixed-Mode Chromatography Resin to remove moderate to high levels of aggregates during downstream processing of therapeutic antibodies in flow-through mode. As a mixed-mode, weak cation-exchange resin, POROS Caprylate resin effectively removes other impurities, such as host cell proteins and a wide range of high molecular weight species. RoboColumns are small chromatography columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms.
Features of POROS Caprylate Mixed-Mode resin include:
- Selective capture of aggregates and hydrophobic high-molecular-weight impurities
- Flow-through design that helps save buffer and material costs
- Excellent scalability and high productivity
- Wide operational range
- Non-animal-derived
POROS Caprylate Mixed-Mode resin (Cat. No. A51051) operates in flow-through mode, which helps reduce costs, especially when working with more engineered monoclonal antibody derivatives. The unique selectivity of the caprylate ligand contributes to a high yield of the targeted monomer while using established cation exchange resin buffer systems. POROS Caprylate Mixed-Mode resin is manufactured using a synthetic manufacturing process that is free of animal components.
Main characteristics
- Matrix: cross-linked poly(styrene-divinylbenzene) POROS beads
- Average particle size: 50 μm
- Ligand: octanoic acid
- Binding capacity: >20 g lysozyme/L resin
- Typical buffers: typical cation exchange buffers, pH 4.5–7.0, conductivity 10–30 mS/cm
- Clean-in-place: 0.5 M NaOH
- Formulation buffer: 0.1 M NaOH
- Mechanical resistance: 100 bar (10 MPa, 1450 psi)
Other POROS chromatography resins are also available in this column format. All POROS high performance chromatography resins have a unique pore structure that provides excellent mass transport for high capacity and resolution over a wide range of flow rates.
RoboColumn™ is a registered trademark of ATOLL GmbH.
Specifications
Specifications
| Max. Pressure | 8 bar |
| Pore Size | Not Applicable |
| Length (Metric) | 3 cm |
| Quantity | 1 Strip, 8 Columns |
| Particle Size | 50 μm |
| For Use With (Equipment) | Robotic Fluid Handlers |
| Type | Mixed Mode Column |
| Diameter (Metric) Inner | 0.5 cm |
| Content And Storage | Store at 2°C to 30°C. Do not freeze. |
| Product Line | POROS |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.