missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ GoPure™ AAVX Column

Product Code. 32045529 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric):
2.5 cm
5 cm
8 cm
10 cm
Length (Metric):
5 cm
10 cm
Bed Volume:
25 mL
50 mL
100 mL
200 mL
250 mL
390 mL
500 mL
785 mL
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) Bed Volume Unit Size
32045529 10 cm 10 cm 785 mL Each
32044809 2.5 cm 5 cm 25 mL Each
32044714 2.5 cm 10 cm 50 mL Each
32044605 5 cm 5 cm 100 mL Each
32044584 5 cm 10 cm 200 mL Each
32045166 8 cm 5 cm 250 mL Each
32045206 8 cm 10 cm 500 mL Each
32045363 10 cm 5 cm 390 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 32045529 Supplier Thermo Scientific™ Supplier No. A65891
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

POROS GoPure AAVX chromatography columns are ready-to-use and designed specifically for use in clinical and commercial manufacturing. POROS GoPure AAVX columns are prepared and packed in an ISO Class 7 environment where all product contact materials meet USP <88> Class VI standards.

POROS GoPure AAVX chromatography columns are ready-to-use and designed specifically for use in clinical and commercial manufacturing. POROS GoPure AAVX columns are prepared and packed in an ISO Class 7 environment where all product contact materials meet USP <88> Class VI standards. Each column is accompanied by column qualification documentation including bioburden and endotoxin. Packed with POROS™ CaptureSelect™ AAVX affinity resin, POROS GoPure AAVX columns offer high specificity and capacity for a broad range of AAV viral vector serotypes over a wide range of flow rates.

  • Pressure rating (bar/psi): 4/58
  • Operating temperature: 2°C to 30°C
  • Bed support size: 12 μm
  • Inlet/outlet internal diameter: 2.00 mm (2.5 cm), 3.45 mm (5 cm), 4.57 mm (8 cm), 6.35 mm (10 cm)
  • Inlet/outlet port connections: 3/4 inch mini tri-clamps
  • Includes a 30 mL resin side sample for incoming QC

Features of POROS CaptureSelect AAVX resin include one-step AAV purification from crude material with high purity, with high specificity and capacity, improving yield by reducing process steps. Free of animal-origin components, the CaptureSelect AAVX ligand is a 12 to 15 kDa single-domain antibody fragment produced in the yeast Saccharomyces cerevisiae.

Specifications

Bed Volume 785 mL
Diameter (Metric) 10 cm
Length (Metric) 10 cm
Quantity 1 Ea.
Length (English) 4 in.

For research and development use and further manufacturing in support of FDA-regulated end uses. Not for diagnostic use or direct administration to human or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.