missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ GoPure™ Caprylate Pre-packed Column, 0.8 x 10 cm, 5 mL

Product Code. 30019301 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 column
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30019301 1 column Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30019301 Supplier Thermo Scientific™ Supplier No. A51055
  • ONLINE EXCLUSIVE PRICE
    933.00 €
    896.00 € / Pack of 1
    Save 37.00 €
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ POROS™ GoPure™ pre-packed columns are intended for the screening of POROS chromatography resins or for small scale purification.

POROS GoPure pre-packed columns are intended for the screening of POROS chromatography resins or for small scale purification.

These pre-packed columns are ready to connect and have the following features:

  • Packed with POROS chromatography resins that offer superior performance over a wide range of flow rates
  • 1-mL and 5-mL formats provide a uniform screening platform for process development
  • Convenient, ready-to-run columns that simplify and speed up POROS resin evaluations and enable small scale purification
  • Direct connection to standard chromatography systems using 1/16 in., 10-32 fittings

POROS Caprylate Mixed-Mode Chromatography Resin (Cat. No. A51051) is designed to remove moderate to high levels of aggregates during downstream processing of therapeutic antibodies in flow-through mode. As a mixed-mode, weak cation-exchange resin, POROS Caprylate resin effectively removes other impurities, such as host cell proteins and a wide range of high molecular weight species.

Features of POROS Caprylate Mixed-Mode resin include:

  • Selective capture of aggregates and hydrophobic high-molecular-weight impurities
  • Flow-through design that helps save buffer and material costs
  • Excellent scalability and high productivity
  • Wide operational range
  • Non-animal-derived

POROS Caprylate Mixed-Mode resin operates in flow-through mode which helps reduce costs, especially when working with more engineered monoclonal antibody derivatives. The unique selectivity of the caprylate ligand contributes to a high yield of the targeted monomer while using established cation exchange resin buffer systems. POROS Caprylate Mixed-Mode resin is manufactured using a synthetic manufacturing process that is free of animal components.

Main characteristics

  • Matrix: cross-linked poly(styrene-divinylbenzene) POROS beads
  • Average particle size: 50 μm
  • Ligand: octanoic acid
  • Binding capacity: >20 g lysozyme/L resin
  • Typical buffers: typical cation exchange buffers, pH 4.5–7.0, conductivity 10–30 mS/cm
  • Clean-in-place: 0.5 M NaOH
  • Formulation buffer: 0.1 M NaOH
  • Mechanical resistance: 100 bar (10 MPa, 1450 psi)

Bulk resin is available and these columns come with a regulatory support package. Worldwide technical support is also available.

Other POROS chromatography resins are also available in this column format. All POROS high performance chromatography resins have a unique pore structure that provides excellent mass transport for high capacity and resolution over a wide range of flow rates.

Specifications

Max. Pressure 30 bar
Pore Size Not Applicable
Length (Metric) 10 cm
Quantity 1 column
Particle Size 50 μm
For Use With (Equipment) Chromatography Systems
Type Mixed Mode Column
Diameter (Metric) Inner 0.8 cm
Content And Storage Store at 2°C to 30°C.
Product Line POROS GoPure
Ligand Type Caprylic Acid
Matrix Cross-linked poly(styrene-divinylbenzene) POROS beads
Target Monoclonal Antibodies
Column Format Pre-packed
Bed Volume 5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.