missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ MabCapture™ A Affinity Chromatography Resin

Product Code. 18426418 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
1 L
Unit Size:
1 L
10mL
250mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426418 10 mL 10mL
18426668 50 mL 50mL
18456518 250 mL 250mL
18416678 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18426418 Supplier Thermo Scientific™ Supplier No. 4374732
  • ONLINE EXCLUSIVE PRICE
    595.00 €
    572.00 € / 10mL
    Save 23.00 €
    3% Off
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific™ POROS™ MabCapture™ A Media is a new design in Protein A chromatography resins technology, combining improvements made in bead and chemistry technology.

POROS MabCapture A Media is a new design in Protein A chromatography resins technology, combining improvements made in bead and chemistry technology. These advances provide a monoclonal antibody purification resins that not only demonstrates the highest dynamic binding performance at low flow rates but also one that can maintain superior binding capacity at high linear velocities where other chromatography resins products cannot operate.

With POROS MabCapture affinity chromatography resins you can:

  • Obtain the highest dynamic binding capacity for monoclonal antibodies vs. conventional agarose or silica based Protein A medias
  • Maintain high dynamic binding capacity at flow rates over 700 cm/hr
  • Design processes with greater flexibility (shorter bed heights, faster flow rates or both) due to the improvements made to the antibody binding efficiency
  • Clean and sanitize with standard sodium hydroxide agents utilized for other chromatography products
  • Recognize major increases to process productivity and reduce the downstream bottleneck

The development of humanized monoclonal antibodies has continued to fuel the growth of the biotherapeutic market. Addressing the therapeutic demand for monoclonal antibodies has lead to major improvements to the cell based expression systems that are used to manufacture these proteins. Consequently, these improvements have moved the processing bottleneck from upstream cell expression to the downstream purification process, as the purification operations struggle to keep up with the large quantities of antibodies that need to be manufactured.

Protein A chromatography, which is used as the primary capture chromatography step in most antibody process platforms, is generally regarded as the major process bottleneck due to the slow flow rates and long residence times required for the effective operation of conventional Protein A chromatography medias.The introduction of POROS MabCapture chromatography resins is aimed at cracking open the downstream bottleneck and providing the capacity, efficiency and flexibility needed to make improvements to the downstream purification platform resulting in major increases to process productivity and antibody manufacturing.

Specifications

Content And Storage Store refrigerated at 2°C to 8°C.
Form Liquid Suspension
Product Line POROS
Quantity 10 mL
Type Bulk Resin
Stationary Phase Protein A

Pharmaceutical Grade Reagent. For Manufacturing and Laboratory Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.