missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ MabCapture™ A RoboColumn™ Pre-packed columns

Product Code. 18486608 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μL Column
600 μL Column
Unit Size:
200µL
600µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18486608 600 μL Column 600µL
18436678 200 μL Column 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18486608 Supplier Thermo Scientific™ Supplier No. A30726
  • ONLINE EXCLUSIVE PRICE
    1170.00 €
    1124.00 € / 600µL
    Save 46.00 €
    3% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ RoboColumn™ chromatography products are small columns provided in 8-column strips.

RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS MabCapture A affinity resin, which is suitable for monoclonal antibody large-scale downstream bioprocessing.

POROS MabCapture A is a 50-micron affinity resin that combines improvements made in bead and chemistry technology. These advances provide a monoclonal antibody purification resin that not only demonstrates the highest dynamic binding performance at low flow rates, but also one that can maintain superior binding capacity at high linear velocities where other chromatography resins products cannot operate.

Features of the resin:

  • Obtain the highest dynamic binding capacity for monoclonal antibodies vs. conventional agarose or silica based Protein A medias
  • Maintain high dynamic binding capacity at flow rates over 700 cm/hr
  • Design processes with greater flexibility (shorter bed heights, faster flow rates or both) due to the improvements made to the antibody binding efficiency
  • Clean and sanitize with standard sodium hydroxide agents utilized for other chromatography products

RoboColumn™ is a registered trademark of ATOLL GmbH.

Specifications

Product Type Column
Product Line MABCAPTURE, POROS
Column Type Affinity
Format Chromatography Cartridge
Stationary Phase Protein A
Purification Target Protein
Content And Storage Store at 2°C to 8°C.
Quantity 600 μL Column
Shipping Condition Wet Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.