missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ XQ RoboColumn™ Pre-packed columns

Product Code. 18436418 Shop All Thermo Scientific Products
Change view
Click to view available options
Column Capacity:
200 μL
600 μL
Unit Size:
200µL
600µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Column Capacity unitSize
18436418 200 μL 200µL
18446448 600 μL 600µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18436418 Supplier Thermo Scientific™ Supplier No. A30719
  • ONLINE EXCLUSIVE PRICE
    270.00 €
    258.00 € / 200µL
    Save 12.00 €
    4% Off
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific™ RoboColumn™ chromatography products are small columns provided in 8-column strips.

RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS XQ resin, a 50-micron strong anion-exchange resin suitable for low-pressure applications, including large-scale downstream bioprocessing.

POROS XQ is a high-capacity, high-resolution, salt-tolerant resin that that enables >140 mg/mL dynamic binding capacity in the presence of up to 6.6 mS/cm NaCl, while delivering exceptional separation performance. A major improvement to the performance of traditional anion exchangers, POROS XQ resin sets the standard for strong anion exchange chromatography in both bind and elute or flow-through modes of operation.

Features of the resin:

  • High dynamic binding capacity leading to reduced column size, smaller footprint, decreased water and buffer usage, and reduced cycling
  • High resolution supporting improved product yield and purity, ultimately improving the cost of goods for the molecule being produced
  • Robust salt tolerance allowing enhanced process design flexibility and streamlining of process flow
  • Low operating back pressure and linear pressure versus flow responses driving flexible scalability
  • Rigid polymeric bead with covalent surface chemistry for easier handling and packing, and superior physical and chemical stability enabling a more robust downstream purification process

RoboColumn™ is a registered trademark of ATOLL GmbH.

Specifications

Flow Rate 16 to 1000 cm/hr.
Column Capacity 200 μL
Quantity 200 μL
Content And Storage Store at 2°C to 8°C or at room temperature.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.