Learn More
Description
RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS XQ resin, a 50-micron strong anion-exchange resin suitable for low-pressure applications, including large-scale downstream bioprocessing.
POROS XQ is a high-capacity, high-resolution, salt-tolerant resin that that enables >140 mg/mL dynamic binding capacity in the presence of up to 6.6 mS/cm NaCl, while delivering exceptional separation performance. A major improvement to the performance of traditional anion exchangers, POROS XQ resin sets the standard for strong anion exchange chromatography in both bind and elute or flow-through modes of operation.
Features of the resin:
- High dynamic binding capacity leading to reduced column size, smaller footprint, decreased water and buffer usage, and reduced cycling
- High resolution supporting improved product yield and purity, ultimately improving the cost of goods for the molecule being produced
- Robust salt tolerance allowing enhanced process design flexibility and streamlining of process flow
- Low operating back pressure and linear pressure versus flow responses driving flexible scalability
- Rigid polymeric bead with covalent surface chemistry for easier handling and packing, and superior physical and chemical stability enabling a more robust downstream purification process
RoboColumn™ is a registered trademark of ATOLL GmbH.
Specifications
Specifications
| Flow Rate | 16 to 1000 cm/hr. |
| Pore Size | Not Applicable |
| Column Capacity | 600 μL |
| Length (Metric) | 3 cm |
| Quantity | 1 Strip, 8 Columns |
| Particle Size | 50 μm |
| For Use With (Equipment) | Robotic Fluid Handlers |
| Type | Anion Exchange Column |
| Diameter (Metric) Inner | 0.5 cm |
| Content And Storage | Store at 2°C to 30°C. Do not freeze. |
| Show More |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.