Learn More
ROBU VitraPOR™ Porous Glass Beads
Substrate for cell cultivation.
Brand: ROBU VitraPOR™ 80080
Additional Details : Weight : 1.09250kg
Description
- Pure, binder-free USP-class I borosilicate glass 3.3
- The special process uses no corrosives and produces a porous structure with smooth, yet large surface areas
- The glass is completely bio-compatible and its high electrical surface charge (Zeta-Potential) helps to stimulate cell growth
Specifications
Borosilicate Glass (3.3) | |
1.5 mm | |
Pore Size: 2 | |
1 Ea. |