missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Porous Plate TransDNA

Product Code. 11405753 Shop All MP Biomedicals Products
Change view
Click to view available options
Type:
Plate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11405753 Plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11405753 Supplier MP Biomedicals Supplier No. 1121008401
  • ONLINE EXCLUSIVE PRICE
    627.00 €
    594.30 € / Pack of 1
    Save 32.70 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for use with vacuum blotters to enable efficient and consistent transfer performance.

  • Replacement part for vacuum blotter ensuring smooth and accurate operation
  • Used during the transfer process to support high-quality blotting results
  • Guarantees perfect transfer in less than one hour without additional handling
  • Delivers nearly 100% transfer efficiency for dependable and reproducible outcomes
  • Usage Statement: Unless specified otherwise, MP Biomedical’s products are for research or further manufacturing use only, not for direct human use

Specifications

For Use With (Equipment) Vacuum Blotter
Type Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.