missing translation for 'onlineSavingsMsg'
Learn More

Porphobilinogen monohydrate, MP Biomedicals™

Product Code. 11493702 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493702 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11493702 Supplier MP Biomedicals Supplier No. 0215633610
  • ONLINE EXCLUSIVE PRICE
    501.00 €
    474.60 € / 10mg
    Save 26.40 €
    5% Off
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 487-90-1
Color White
Molecular Formula C10H14N2O4
Quantity 10 mg
Synonym porphobilinogen, unii-74khc72qxk, 3-5-aminomethyl-4-carboxymethyl-1h-pyrrol-3-yl propanoic acid, 1h-pyrrole-3-propanoic acid, 5-aminomethyl-4-carboxymethyl, 2-aminomethylpyrrol-3-acetic acid 4-propionic acid, 74khc72qxk, pbg, 5-aminomethyl-4-carboxymethyl-1h-pyrrole-3-propanoic acid, 5-aminomethyl-4-carboxymethyl-1h-pyrrole-3-propionic acid, 5-aminomethyl-4-carboxymethyl-pyrrole-3-propionic acid
InChI Key QSHWIQZFGQKFMA-UHFFFAOYSA-N
SMILES C1=C(C(=C(N1)CN)CC(=O)O)CCC(=O)O
IUPAC Name 3-[5-(aminomethyl)-4-(carboxymethyl)-1H-pyrrol-3-yl]propanoic acid
Molecular Weight (g/mol) 226.232
PubChem CID 1021
ChEBI CHEBI:17381
Formula Weight 244.3
Physical Form Powder
Chemical Name or Material Porphobilinogen monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Porphobilinogen Monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Porphobilinogen Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.