missing translation for 'onlineSavingsMsg'
Learn More

Porphobilinogen, TRC

Product Code. 30412140
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412140 10 mg 10mg
30413165 5 mg -
30410490 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30412140 Supplier Toronto Research Chemicals Supplier No. TRCP68950010MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 487-90-1
Molecular Formula C10H14N2O4
Molecular Weight (g/mol) 226.23
Synonym 5-(Aminomethyl)-4-(carboxymethyl)-1H-pyrrole-3-propanoic Acid, PBG, 3-[5-(Aminomethyl)-4-(carboxymethyl)-1H-pyrrol-3-yl]propanoic Acid hydrate
IUPAC Name 3-[5-(aminomethyl)-4-(carboxymethyl)-1H-pyrrol-3-yl]propanoic acid
SMILES NCc1[nH]cc(CCC(=O)O)c1CC(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H14N2O4/c11-4-8-7(3-10(15)16)6(5-12-8)1-2-9(13)14/h5,12H,1-4,11H2,(H,13,14)(H,15,16)
Formula Weight 226.0954
Percent Purity >95%
Chemical Name or Material Porphobilinogen
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.