missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Portable Waterproof pH/EC/TDS Meters

Product Code. 10142321
Change view
Click to view available options
For Use With (Application):
High-Range Detection
Range (Conductivity):
0.00 to 20.00mS/cm
Range (TDS):
0.00 to 10.00ppt (g/L)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Range (Conductivity) Range (TDS) unitSize
10142321 High-Range Detection 0.00 to 20.00mS/cm 0.00 to 10.00ppt (g/L) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10142321 Supplier Hanna Instruments™ Supplier No. HI991301
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Designed to measure pH, conductivity (EC), total dissolved solids (TDS), and temperature with a single probe. Hanna Instruments™ Portable Waterproof pH/EC/TDS Meters are available for high range and low range measurement.

This multi-parameter meter takes readings simply and quickly, and it fits in the palm of your hand. From setup to calibration all operations are performed with just two buttons. It features a clear, extra-large LCD display and easy-access rotary knobs to effect calibration changes. A single pre-amplified probe is used for all parameters.

  • Waterproof and rated for IP 67 conditions
  • Easy to clean and maintain
  • Multi-level LCD display
  • On-screen tutorial messages for calibration and setup
  • Automatic Temperature Compensation
  • Automatic one or two-point pH calibration
  • BEPS: Alerts the user in the event that low battery power could adversely affect the readings
  • Battery % displayed on startup
  • Compact, heavy duty, and waterproof
  • Preprogrammed buffer recognition values, TDS factors and temperature coefficients

Specifications

Includes HI 1288 probe, batteries, 1m cable, rugged carrying case and instructions
Accuracy (Temperature) ±0.5°C/±1°F
Accuracy (pH) ±0.01
For Use With (Application) High-Range Detection
Calibration Automatic
Range (pH) 0 to 14
Resolution (pH) 0.01
Temperature Range (Metric) 0°C to 60.0°C
Temperature Range (English) 32.0°F to 140.0°F
Resolution (Temperature) 0.1°C/F
Display Type LCD
Certifications/Compliance IP 67
Dimensions (L x W x H) 150 x 80 x 36 mm (5.9 x 3.1 x 1.4 in.)
Weight (Metric) 205 g
Weight (English) 7.2 oz.
Warranty 2 Years (6 Months Probe)
Keypad Two-button
Accuracy (Conductivity) ±2% F.S.
Material PVC
Probe Type HI1288 PVC body, pre-amplified multiparameter probe with internal temperature sensor, DIN connector and 1m cable
Range (Conductivity) 0.00 to 20.00mS/cm
Range (TDS) 0.00 to 10.00ppt (g/L)
Resolution TDS Conversion Factor: 0.45 to 1.00
Resolution (Conductivity) 0.01mS/cm
Resolution (TDS) 0.01ppt (g/L)
Detection Range 0 to 14 pH
Temperature Compensation pH: automatic; EC/TDS: automatic with β adjustable from 0.0 to 2.4%/°C
Battery Type 1.5V (3) AAA, Auto-Off after 8 min. non-use
Type Portable pH/EC/TDS/Temperature Meter
Battery Life 500 hr.
Accuracy (TDS) ±2% full scale
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.