Learn More
Hach Lange™ Portable Turbidimeter
Product Code. 12311693
Change view
Click to view available options
Light Source:
LED
Unit Size:
Each
Product Code. Light Source Unit Size
12311693 LED Each
1 options
Product Code. 12311693 Supplier Hach Lange™ Supplier No. 2100QIS01
Spotlight Offers
Use promo code 28038
View all promotions
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Portable Turbidimeter offers unsurpassed ease of use and accuracy in turbidity measurement.
  • With easy-to-follow on-screen instructions, manuals or memorization are not needed for routine calibrations.
  • The Rapidly Settling Turbidity mode allows for accurate measurements even with the most difficult-to-measure settling samples.
  • The rugged design makes this the instrument of choice for the toughest environments.
TRUSTED_SUSTAINABILITY
Specifications
Light Source LED
Description Portable Turbidimeter
Accuracy ±2%