missing translation for 'onlineSavingsMsg'
Learn More

PSF Progressive Safety™ Portwest™ Antistatic Laced Janitorial Boot

Product Code. 15971662
Change view
Click to view available options
Size:
4
6
7
8
9
10
11
12
Unit Size:
1 pair
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15971662 4 1 pair
15619588 6 1 pair
15689578 7 1 pair
15699578 8 1 pair
15609588 9 1 pair
15659578 10 1 pair
15669578 11 1 pair
15679578 12 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15971662 Supplier PSF Progressive Safety™ Supplier No. FW884
  • ONLINE EXCLUSIVE PRICE
    60.38 €
    54.46 € / 1 pairs
    Save 5.92 €
    9% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Providing water resistant safety at an affordable price.

  • All safety footwear is designed with both comfort and durability in mind
  • Conforms to EN ISO 20345:2011 for 200 joules protection
  • Water resistant upper
  • Antistatic and antislip dual density outsole
  • Laced version ideal for janitorial work
  • Antibacterial lining
  • CE certified

Specifications

Certifications/Compliance EN ISO 20345:2011
Color White
Design Laced boot
For Use With (Application) Janitorial work
Material Microfiber
Size 4
Sole Features Energy absorbing, slip-resistant
Resistant To Water, oil, slip-resistant
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.