Learn More
Globe Scientific PosiStop LTS-Compatible Pipette Tips, Empty Racks
Product Code. 31923694
Change view
Click to view available options
Holds:
Pipette Tips (1000 μL)
Pipette Tips (20 μL)
Pipette Tips (200 μL)
Unit Size:
Pack of 10
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
31923694 Pipette Tips (20 μL) Pack of 10
31923664 Pipette Tips (200 μL) Pack of 10
31924095 Pipette Tips (1000 μL) Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31923694 Supplier Globe Scientific Supplier No. 154405
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
Empty racks for PosiStop pipette tips in 20 μL (red tray), 200 μL (green tray), and 1000 μL (blue tray) sizes. Durable, reusable, and color-coded for quick volume ID and efficient lab organization. Ideal for refilling and maintaining clean, organized tip storage.
  • Empty racks for 20 μL, 200 μL, and 1000 μL PosiStop pipette tips
  • Color-coded trays: red (20 μL), green (200 μL), blue (1000 μL)
  • Durable and reusable for efficient lab workflows
  • Ideal for refilling and organized tip storage
Specifications
Autoclavable Autoclavable
Color Natural
Holds Pipette Tips (20 μL)
Material Polypropylene
Positions 96
Shape Rectangular
Stackable Yes
Sterility Non-sterile
Product Type Empty Rack
Array 12 x 8