Learn More
Gilson™ Position Rack
Product Code. 16686125 Shop All Gilson Products
Change view
Click to view available options
For Use With (Equipment):
5 mL Vial (12 x 75/100 mm)
9 mL Vial (13 x 100 mm)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16686125 5 mL Vial (12 x 75/100 mm) Each
16666125 9 mL Vial (13 x 100 mm) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16686125 Supplier Gilson™ Supplier No. 170414
  • ONLINE EXCLUSIVE PRICE
    1495.00 €
    1110.61 € / Pack of 1
    Save 384.39 €
    25% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
These racks are for use with the FC203B and FC204 Fraction Collectors.
  • HPLC Purification Technique
  • Fraction Collector Racks
Specifications
Description Code 14
For Use With (Equipment) 5 mL Vial (12 x 75/100 mm)
No. of Positions 128