missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies Positioning Plate For LB 9515

Product Code. 31777372 Shop All Berthold Technologies Products
Change view
Click to view available options
For Use With (Application):
Luminescence QC
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
31777372 Luminescence QC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31777372 Supplier Berthold Technologies Supplier No. 53613
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in the NightSHADE and NightOWL imaging systems to keep the calibration plate in the right position.

  • Luminescence Test Plate LB 9515 is used to verify performance of Berthold luminometers and imaging systems
  • The positioning plate is used with NightSHADE and NightOWL imaging systems
  • Ensures correct placement of the calibration plate during image acquisition

Specifications

Type Positioning Plate
Color Black
For Use With (Application) Luminescence QC
For Use With (Equipment) NightSHADE evo LB 985 in vivo Imaging System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.