missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-chlorophenyltrifluoroborate, 96%, Thermo Scientific™

Product Code. 15499987
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15499987

Brand: Thermo Scientific Alfa Aesar H53231.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Potassium 4-chlorophenyltrifluoroborate is involved in Suzuki Cross-Coupling reaction as potent boronic acid surrogates.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents and acids. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 661465-44-7
Molecular Formula C6H4BClF3K
Molecular Weight (g/mol) 218.452
MDL Number MFCD04115770
InChI Key GXBSKMLVYJPMRT-UHFFFAOYSA-N
Synonym potassium 4-chlorophenyltrifluoroborate, potassium 4-chlorophenyl trifluoroborate, potassium 4-chlorophenyl trifluoroboranuide, pubchem11348, potassium 4-chlorophenyl trifluoro borate 1-, potassium ion 4-chlorophenyl trifluoroboranuide
PubChem CID 23682053
IUPAC Name potassium;(4-chlorophenyl)-trifluoroboranuide
SMILES [B-](C1=CC=C(C=C1)Cl)(F)(F)F.[K+]

Specifications

Melting Point 293°C to 295°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 218.45
Percent Purity 96%
Chemical Name or Material Potassium 4-chlorophenyltrifluoroborate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.