missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-(methoxycarbonyl)phenyltrifluoroborate, 96%, Thermo Scientific™

Product Code. 15429925
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15429925 1 g 1g
15439925 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15429925 Supplier Thermo Scientific Alfa Aesar Supplier No. H52588.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Potassium 4-(methoxycarbonyl)phenyltrifluoroborate is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 705254-34-8
Molecular Formula C8H7BF3KO2
Molecular Weight (g/mol) 242.05
MDL Number MFCD04115741
InChI Key IQYSKWRPRJKKAG-UHFFFAOYSA-N
Synonym potassium 4-methoxycarbonylphenyl trifluoroborate, potassium trifluoro 4-methoxycarbonyl phenyl boranuide, potassium trifluoro 4-methoxycarbonyl phenyl borate, c8h7bf3o2.k, potassium 4-methoxycarbonyl trifluoroborate, trifluoro 4-methoxycarbonyl phenyl potassioboron v, potassium trifluoro-4-methoxycarbonylphenyl boranuide, potassium trifluoro 4-methoxycarbonyl phenyl borate 1-, potassium ion trifluoro 4-methoxycarbonyl phenyl boranuide, potassium tris fluoranyl-4-methoxycarbonylphenyl boranuide
PubChem CID 23716869
IUPAC Name potassium;trifluoro-(4-methoxycarbonylphenyl)boranuide
SMILES [K+].COC(=O)C1=CC=C(C=C1)[B-](F)(F)F

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 242.05
Percent Purity 96%
Chemical Name or Material Potassium 4-(methoxycarbonyl)phenyltrifluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.