missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-methoxyphenyltrifluoroborate, 95%

Product Code. 11367179
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367179 5 g 5g
11357179 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367179 Supplier Thermo Scientific Alfa Aesar Supplier No. H27384.06
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 192863-36-8 | C7H7BF3KO | 214.036 g/mol

It is applied in Suzuki Cross-Coupling using organotrifluoroborates as potent boronic acid surrogates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in Suzuki Cross-Coupling using organotrifluoroborates as potent boronic acid surrogates.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192863-36-8
Molecular Formula C7H7BF3KO
Molecular Weight (g/mol) 214.036
MDL Number MFCD04112717
InChI Key XNYMCUFKZHRYID-UHFFFAOYSA-N
Synonym potassium trifluoro 4-methoxyphenyl borate, potassium 4-methoxyphenyl trifluoroborate, potassium 4-methoxyphenyltrifluoroborate, potassium trifluoro 4-methoxyphenyl boranuide, pubchem11347, amtb103, potassum 4-methoxyphenyl trfluoroborate, trifluoro 4-methoxyphenyl potassioboron v, potassium trifluoro-4-methoxyphenyl boranuide
PubChem CID 23683762
IUPAC Name potassium;trifluoro-(4-methoxyphenyl)boranuide
SMILES [B-](C1=CC=C(C=C1)OC)(F)(F)F.[K+]

Specifications

Melting Point >300°C
Quantity 5 g
Formula Weight 214.04
Percent Purity 95%
Chemical Name or Material Potassium 4-methoxyphenyltrifluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.