Learn More
Melford Potassium Acetate
Description
In research laboratories, potassium acetate is utilized for various purposes.
Potassium acetate is also used in protein purification techniques, such as ammonium sulfate precipitation, to selectively precipitate proteins from solution based on their solubility characteristics.
Furthermore, potassium acetate is utilized as a buffering agent in biochemical assays and cell culture media. Its buffering capacity helps maintain a stable pH environment, which is critical for many enzymatic reactions and cell culture processes
Specifications
Specifications
| Chemical Name or Material | Potassium Acetate |
| CAS | 127-08-2 |
| Quantity | 2.5 kg |
| Molecular Formula | C2H3O2K |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.