missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acetate, ≥99.0%, ACS Reagent, Solstice

Product Code. 15622340
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15622340 100 g Poly Bottle 100g
15642340 500 g Poly Bottle 500g
15632340 2.5 kg Poly Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15622340 Supplier Honeywell-Fluka Supplier No. 236497100G
  • ONLINE EXCLUSIVE PRICE
    53.00 €
    47.45 € / 100g
    Save 5.55 €
    10% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Tekniske data

Melting Point 292°C
Physical Form Solid
Quantity 100 g
Packaging Poly Bottle
Density 1.57g/cu.cm at 25°C
Linear Formula CH3COOK
UN Number NONH for all modes of transport
Beilstein 3595449
Solubility Information Soluble in water, 98.2g/L at 20°C
Formula Weight 98.14g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Chemical Name or Material Potassium Acetate
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.