missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acetate, BAKER ANALYZED™ ACS Reagent, J.T. Baker™

Product Code. 15568064 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly bottle
Unit Size:
2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15568064 2.5 kg Poly bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15568064 Supplier Avantor J.T.Baker Supplier No. 291205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 127-08-2
Physical Form Crystals
Quantity 2.5 kg
Molecular Formula C2H3KO2
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
SMILES CC(=O)[O-].[K+]
IUPAC Name potassium;acetate
Molecular Weight (g/mol) 98.142
PubChem CID 517044
ChEBI CHEBI:32029
Formula Weight 98.14
Grade Meets ACS reagent chemical requirements
Packaging Poly bottle
Specific Gravity 1L = 1.57kg
Chemical Name or Material Potassium Acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.