missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acetate, (Crystalline/ACS Reagent Grade), MP Biomedicals

Product Code. 11480542 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480542 1 kg 1kg
11460542 100 g 100g
11470542 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480542 Supplier MP Biomedicals Supplier No. 0219142591
 Restricted Item
Estimated Shipment: 24-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

Physical Form Crystalline
Quantity 1 kg
Formula Weight 98.14
Percent Purity 99.72%
Grade ACS Reagent
Chemical Name or Material Potassium Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.