missing translation for 'onlineSavingsMsg'
Learn More

Potassium benzoate, 99%

Product Code. 11494693
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494693 1000 g 1000g
11404703 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494693 Supplier Thermo Scientific Alfa Aesar Supplier No. A11610.0B
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 582-25-2 | C7H5KO2 | 160.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Potassium Benzoate is manufactured primarily for food and beverage use. It is a chemical preservative, which in very low concentrations inhibits the activity of the microorganisms. It is used in carbonated beverages. The shelf life of un-pasteurized cider can be greatly extended by adding potassium benzoate. It is also used as the whistle in many fireworks.

Solubility
Soluble in ethanol, slightly soluble in methanol, insoluble in ether.

Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 582-25-2
Molecular Formula C7H5KO2
Molecular Weight (g/mol) 160.213
MDL Number MFCD00013061
InChI Key XAEFZNCEHLXOMS-UHFFFAOYSA-M
Synonym potassium benzoate, benzoic acid, potassium salt, potassium salt, unii-763yqn2k7k, potassium benzoate nf, benzoic acid potassium salt, potassium ion benzoate, acmc-20ajv6, dsstox_cid_7219
PubChem CID 23661960
IUPAC Name potassium;benzoate
SMILES C1=CC=C(C=C1)C(=O)[O-].[K+]

Specifications

Melting Point >300°C
Assay Percent Range 99%
Quantity 1000 g
Beilstein 3719165
Sensitivity Hygroscopic
Merck Index 14,1091
Solubility Information Soluble in ethanol,slightly soluble in methanol,insoluble in ether.
Formula Weight 160.22
Percent Purity 99%
Chemical Name or Material Potassium benzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.