missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bicarbonate, ACS Reagent, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
Beskrivelse
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Tekniske data
Tekniske data
| Physical Form | Powder, Crystals and/or Granules |
| Quantity | 100 g |
| Linear Formula | KHCO3 |
| UN Number | NONH for all modes of transport |
| Beilstein | 4535309 |
| Formula Weight | 100.12g/mol |
| Percent Purity | 99% |
| Grade | ACS Reagent |
| Chemical Name or Material | Potassium Bicarbonate |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion