missing translation for 'onlineSavingsMsg'
Learn More

Potassium bicarbonate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, E501, 99.5-101.0% (acidimetric), Solstice

Product Code. 15661150
Change view
Click to view available options
Quantity:
1 kg
25 kg
6 x 1 kg
Packaging:
Paper / Poly Bag
Poly Bottle
Unit Size:
1kg
25kg
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15661150 6 x 1 kg Poly Bottle Pack of 6
15641150 1 kg Poly Bottle 1kg
15651150 25 kg Paper / Poly Bag 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15661150 Supplier Honeywell-Fluka Supplier No. 126026X1KG
  • ONLINE EXCLUSIVE PRICE
    235.00 €
    211.75 € / Pack of 6
    Save 23.25 €
    9% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., USP, E501, 99.5-101.0% (acidimetric)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
IUPAC Name potassium;hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

CAS Max % 100.0000%
Physical Form Powder
Quantity 6 x 1 kg
Assay Percent Range 99.5 to 101.0%
Linear Formula KHCO3
UN Number NONH for all modes of transport
Beilstein 4535309
Solubility Information Soluble in water
Formula Weight 100.12
Percent Purity 99.5 to 101.0% (acidimetric)
Grade puriss.
Packaging Poly Bottle
Density 2.17 g/cm3
Chemical Name or Material Potassium bicarbonate, Puriss., meets analytical specification of BP, Ph. Eur., USP, E501, 99.5-101.0% (acidimetric)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.