missing translation for 'onlineSavingsMsg'
Learn More

Potassium Biphthalate, ULTREX™ Ultrapure Reagent, J.T. Baker™

Product Code. 15558314 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15558314 25 g Wide Mouth Amber Glass Bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15558314 Supplier Avantor J.T.Baker Supplier No. 488903
  • ONLINE EXCLUSIVE PRICE
    769.00 €
    719.11 € / Pack of 1
    Save 49.89 €
    6% Off
 Restricted Item
Estimated Shipment: 22-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For Primary Standard and Research Applications

Specifications

CAS 877-24-7
Quantity 25 g
Molecular Formula C8H5KO4
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
IUPAC Name potassium;2-carboxybenzoate
Molecular Weight (g/mol) 204.222
PubChem CID 23676735
Formula Weight 204.22
Grade ULTREX™ Ultrapure Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.64kg
Chemical Name or Material Potassium Biphthalate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.