missing translation for 'onlineSavingsMsg'
Learn More

Potassium bisulfate, 98.5%, for analysis

Product Code. 10378943 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10378943 2.5 kg Glass bottle 2.5kg
10542611 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10378943 Supplier Thermo Scientific Acros Supplier No. 207750025
  • ONLINE EXCLUSIVE PRICE
    185.00 €
    179.00 € / 2.5kg
    Save 6.00 €
    3% Off
View all promotions
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7646-93-7 | HKO4S | 136.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-93-7
Molecular Formula HKO4S
Molecular Weight (g/mol) 136.16
MDL Number MFCD00011404
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
Synonym potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
PubChem CID 516920
IUPAC Name potassium hydrogen sulfate
SMILES [K+].OS([O-])(=O)=O

Specifications

Melting Point 214.0°C
Color White
Physical Form Adhering Crystals
Quantity 2.5 kg
Assay Percent Range 98.50%
Linear Formula KHSO4
Fieser 01,909
Merck Index 15, 7734
Solubility Information Solubility in water: 490g/L (20°C). Other solubilities: decomposes in alcohol
Formula Weight 136.16
Percent Purity 98.5%
Grade Analytical
Packaging Glass bottle
Density 2.32
Chemical Name or Material Potassium bisulfate, c.p.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium bisulfate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.