missing translation for 'onlineSavingsMsg'
Learn More

Potassium bisulfate, Puriss. p.a., ≥99%, Solstice

Product Code. 15623350
Change view
Click to view available options
Quantity:
500 g
1 kg
50 kg
6 x 1 kg
Unit Size:
1kg
500g
50kg
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15623350 6 x 1 kg Pack of 6
15603350 500 g 500g
15693340 1 kg 1kg
15613350 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15623350 Supplier Honeywell-Fluka Supplier No. 312606X1KG
  • ONLINE EXCLUSIVE PRICE
    628.00 €
    567.62 € / Pack of 6
    Save 60.38 €
    9% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a.,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-93-7
Molecular Formula HKO4S
Molecular Weight (g/mol) 136.16
MDL Number MFCD00011404
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
Synonym potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
PubChem CID 516920
IUPAC Name potassium;hydrogen sulfate
SMILES [K+].OS([O-])(=O)=O

Specifications

Physical Form Powder
Quantity 6 x 1 kg
Linear Formula KHSO4
UN Number UN2509
Formula Weight 136.17
Percent Purity ≥99%
Grade puriss. p.a.
Density 2.32g/mL at 25°C
Chemical Name or Material Potassium Bisulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.