missing translation for 'onlineSavingsMsg'
Learn More

Potassium bisulfate, Puriss. p.a., ≥99%, Solstice

Product Code. 16695793
Change view
Click to view available options
25 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16695793 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16695793 Supplier Fluka Supplier No. 3126025KG
 Restricted Item
Estimated Shipment: 08-02-2030
Add to basket
This item is not returnable. View return policy

Specifications

Melting Point 200°C
CAS 7646-93-7
Color Colorless
CAS Max % 1
Quantity 25 kg
Molecular Formula HKO4S
Linear Formula KHSO4
MDL Number MFCD00011404
Synonym Potassium hydrogen sulfate
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
SMILES [K+].OS([O-])(=O)=O
IUPAC Name dipotassium sulfate
Molecular Weight (g/mol) 136.16
Grade ACS
Density 2.36 g/cm3 (20°C)
Chemical Name or Material Potassium bisulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.