missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, 99.92% (metals basis)

Product Code. 11321578
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11321578 100 g 100g
11311578 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11321578 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur 010982.22
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 7758-02-3 | BrK | 119.00 g/mol

Potassium bromide is used in manufacturing of photographic papers, engraving and lithography, in spectroscopy and as an analytical lab reagent. Potassium bromide is employed in antiepileptic medication for dogs. It is utilized in the bromination of alkenes using cerium(IV) ammonium nitrate. It plays an essential role with diacetoxy iodobenzene for bromination of alkenes and alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
IUPAC Name potassium bromide
SMILES [K+].[Br-]

Specifications

Boiling Point 1435°C
Melting Point 730°C
Color Colorless
Physical Form Crystalline
Quantity 100 g
Odor Odorless
Refractive Index 1.559
Sensitivity Hygroscopic
Appearance White Powder/crystalline/beads
Assay Percent Range 99.92% (metals basis)
Density 2.75 g/cm3 at 20°C
Solubility Information Freely soluble in water. Soluble in alcohol and glycerol. Slightly soluble in diethyl ether.
Formula Weight 119.01
Chemical Name or Material Potassium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.