missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beskrivelse
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For IR spectroscopy
Tekniske data
Tekniske data
| Boiling Point | 1435.0°C |
| Melting Point | 734.0°C |
| Color | White |
| Physical Form | Fine Crystalline Powder |
| Quantity | 500 g |
| Assay Percent Range | 99% min. (Argentometry) |
| Linear Formula | KBr |
| Merck Index | 15, 7739 |
| Solubility Information | Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C),1.42g/L ethanol (25°C),217g/L glycerol,4g/L alcohol |
| IUPAC Name | potassium bromide |
| Vis mere |
Sikkerhed og håndtering
Produktidentifikator
- Potassium bromide
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?