missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, 99+%, for spectroscopy, IR grade

Product Code. 10793201 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10793201 100 g Plastic bottle 100g
10318243 25 g Plastic bottle 25g
10003163 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10793201 Supplier Thermo Scientific Acros Supplier No. 206391000
  • ONLINE EXCLUSIVE PRICE
    96.25 €
    90.20 € / 100g
    Save 6.05 €
    6% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7758-02-3 | BrK | 119.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For IR spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-02-3
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

Boiling Point 1435.0°C
Melting Point 734.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 100 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Assay Percent Range 99% min. (Argentometry)
Linear Formula KBr
Merck Index 15, 7739
Solubility Information Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C), 1.42g/L ethanol (25°C), 217g/L glycerol, 4g/L alcohol
IUPAC Name potassium;bromide
Formula Weight 119
Percent Purity ≥99%
Grade Spectroscopy
Chemical Name or Material Potassium Bromide, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Potassium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.