missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, 99+%

Product Code. 11464020
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1000 g
2.5 kg
5000 g
Unit Size:
1000g
100g
2.5kg
250g
5000g
500g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464020 500 g 500g
11454020 100 g 100g
11447807 250 g 250g
11437807 1000 g 1000g
11457807 5000 g 5000g
11474020 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11464020 Supplier Thermo Scientific Alfa Aesar Supplier No. A16339.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7758-02-3 | BrK | 119.00 g/mol

Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.

Solubility
Slightly soluble in alcohol but insoluble in acetone and ethanol.

Notes
Strong oxidizer. Incompatible with combustible materials, organics, reducing agents, aluminum and finely powdered metals
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

Boiling Point 1435°C
Melting Point 730°C
Quantity 500 g
Merck Index 14,7618
Solubility Information Slightly soluble in alcohol but insoluble in acetone and ethanol.
IUPAC Name potassium;bromide
Formula Weight 119.01
Percent Purity ≥99%
Odor Odorless
Refractive Index 1.559
Sensitivity Hygroscopic
Density 2.75
Chemical Name or Material Potassium bromide
Show More Show Less
compliance-icons
Product Identifier
  • Potassium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Potassium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.