missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, ACS, 99% min

Product Code. 11337467
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Unit Size:
1kg
250g
50g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337467 1 kg 1kg
11357467 50 g 50g
11327467 250 g 250g
11347467 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11337467 Supplier Thermo Scientific Alfa Aesar Supplier No. 012612.A1
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Manufacturing of Photographic Papers, Process Engraving and Lithography, Spectroscopy | CAS: 7758-02-3 | BrK | 119.00 g/mol

Used in manufacturing of photographic papers, engraving and lithography, in spectroscopy, and as an analytical lab reagent. Potassium bromide is applied as a veterinary drug, as an antiepileptic medication for dogs. It is utilized for the bromination of alkenes using cerium(IV) ammonium nitrate. It plays an essential role with diacetoxy iodobenzene for the efficient bromination of alkenes and alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

Boiling Point 1435°C
Melting Point 730°C
Color White
Physical Form Crystalline
Quantity 1 kg
Assay Percent Range 99% min.
Solubility Information Freely soluble in water. Soluble in alcohol,glycerol.
IUPAC Name potassium bromide
Formula Weight 119.01
Grade ACS
Odor Odorless
Refractive Index 1.559
Sensitivity Hygroscopic
Appearance White powder/crystalline/beads
Density 2.75
Chemical Name or Material Potassium bromide, For analysis ACS
Show More Show Less
compliance-icons
Product Identifier
  • Potassium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Potassium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.