missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide crystal optic rectangle, 30mm x 15mm x 4mm, polished both sides

Product Code. 15463645
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15463645 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15463645 Supplier Thermo Scientific Alfa Aesar Supplier No. 038457.KT
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7758-02-3 | BrK | 119.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2-3-7758
Quantity 1 Ea.
Molecular Formula BrK
MDL Number MFCD00011358
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES [K+].[Br-]
IUPAC Name potassium;bromide
Molecular Weight (g/mol) 119.00
PubChem CID 253877
ChEBI CHEBI:32030
Chemical Name or Material Potassium bromide crystal optic rectangle, 30mm x 15mm x 4mm, Polished both sides
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.