missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, Premium FTIR Grade

Product Code. 15452847
Change view
Click to view available options
Quantity:
200 Pc.
25 Pc.
50 Pc.
Unit Size:
Pack of 200
Pack of 25
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15452847 200 Pc. Pack of 200
15432847 25 Pc. Pack of 25
15422847 50 Pc. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15452847 Supplier Thermo Scientific Alfa Aesar Supplier No. 047111.KQ
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7758-02-3 | BrK | 119.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
IUPAC Name potassium bromide
SMILES [K+].[Br-]

Spécification

Boiling Point 1435°C
Melting Point 730°C
Physical Form Powder
Quantity 200 Pc.
Odor Odorless
Packaging 0.5g packets
Refractive Index 1.559
Density 2.75 g/mL
Linear Formula KBr
Merck Index 14,7618
Formula Weight 119.01
Grade Premium FTIR
Chemical Name or Material Potassium bromide
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.