missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, puriss., Solstice
BIOPHARMA

Product Code. 15620560
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
25 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
100g
1kg
2.5kg
25kg
500g
Pack of 6
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15620560 100 g 100g
15660560 500 g 500g
15630560 1 kg 1kg
15640560 2.5 kg 2.5kg
15670560 6 x 1 kg Pack of 6
15680560 6 x 2.5 kg Pack of 6
15650560 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15620560 Supplier Honeywell-Fluka Supplier No. 02110100G
  • ONLINE EXCLUSIVE PRICE
    42.00 €
    37.90 € / 100g
    Save 4.10 €
    9% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99.5-100.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES [K+].[Br-]

Specifications

CAS 7758-02-3, 14708-99-7
Physical Form Solid
Quantity 100 g
Linear Formula KBr
UN Number NONH for all modes of transport
IUPAC Name potassium bromide
Formula Weight 119g/mol
Percent Purity 99.5 to 100.5%
Grade puriss.
Packaging Plastic Bottle
Vapor Pressure 0.01mmHg (20°C), 1mmHg (795°C)
Chemical Name or Material Potassium Bromide, Meets analytical specification of Ph. Eur.BP,USP
Show More Show Less
compliance-icons
Product Identifier
  • Potassium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Potassium bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.