missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide solution 1mol/L, Fisher Chemical™

Product Code. 12923574 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12923574 1000 mL Bottle 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12923574 Supplier Fisher Chemical Supplier No. VL 255 1000
  • ONLINE EXCLUSIVE PRICE
    135.00 €
    133.26 € / 1000mL
    Save 1.74 €
    1% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

CAS 7758-02-3, 7732-18-5
Quantity 1000 mL
Molecular Formula BrK
MDL Number MFCD00011358
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES [K+].[Br-]
IUPAC Name potassium bromide
Molecular Weight (g/mol) 119.00
PubChem CID 253877
ChEBI CHEBI:32030
Concentration or Composition (by Analyte or Components) 1M (1N)
Packaging Bottle
Chemical Name or Material Potassium Bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium bromide 1M Volumetric solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P314-Get medical advice/attention if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: potassium bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.