missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromomethyltrifluoroborate, 95%

Product Code. 10515490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10515490 1 g 1g
10008239 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10515490 Supplier Thermo Scientific Acros Supplier No. 431530010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 888711-44-2 | CH2BBrF3K | 200.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 888711-44-2
Molecular Formula CH2BBrF3K
Molecular Weight (g/mol) 200.83
InChI Key AZDFPIRYUOCVCJ-UHFFFAOYSA-N
Synonym potassium bromomethyl trifluoroborate, potassium bromomethyltrifluoroborate, potassium bromomethyl trifluoroboranuide, potassium bromomethyl trifluoro boranuide, potassium bromomethyl trifluoro borate 1-, borate 1-, bromomethyl trifluoro-, potassium 1:1 , t-4, pubchem11564, bromomethyl potassium trifluoroborate, postassium bromomethyltrifluoroborate, trifluoro bromomethyl potassioboron v
PubChem CID 23690312
IUPAC Name potassium;bromomethyl(trifluoro)boranuide
SMILES [B-](CBr)(F)(F)F.[K+]

Specifications

Melting Point 220.0°C
Color White
Physical Form Powder
Quantity 1 g
Assay Percent Range 95%
Formula Weight 200.83
Percent Purity 95%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Potassium bromomethyltrifluoroborate
compliance-icons
Product Identifier
  • Potassium bromomethyltrifluoroborate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.