missing translation for 'onlineSavingsMsg'
Learn More

Potassium (Butoxycarbonothioyl)sulfide, TRC

Product Code. 30417642
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417642 500 mg 500mg
30414493 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30417642 Supplier Toronto Research Chemicals Supplier No. TRCP993585500MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium (Butoxycarbonothioyl)sulfide
CAS 871-58-9
Molecular Formula C5 H9 O S2 . K
Quantity 500 mg
Synonym Carbonic acid','dithio-','O-butyl ester','potassium salt (8CI), Carbonodithioic acid','O-butyl ester','potassium salt (9CI), Xanthic acid','butyl-','potassium salt (6CI), Butyl potassium xanthate, Butyl potassium xanthogenate, O-Butyl potassium dithiocarbonate, Potassium butyl dithiocarbonate, Potassium butyl xanthate, Potassium butylxanthogenate, Potassium n-butylxanthate, Potassium O-butyl carbonodithioate, Potassium O-butyl dithiocarbonate, Potassium O-butyl xanthate, Potassium O-n-butyl xanthate, Carbonodithioic acid','O-butyl ester','potassium salt (1:1) (ACI), Butylxanthic Acid Potassium
InChI Formula InChI=1S/C5H10OS2.K/c1-2-3-4-6-5(7)8;/h2-4H2,1H3,(H,7,8);/q;+1/p-1
SMILES [K+].CCCCOC(=S)[S-]
IUPAC Name potassium;butoxymethanedithioate
Molecular Weight (g/mol) 188.35
Formula Weight 187.9732
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.