missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate Anhydrous, Extra Pure, SLR, Fisher Chemical™

Product Code. 10637612 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
2.5kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10637612 5 kg Plastic keg 5kg
10121440 500 g Plastic powder jar 500g
10558820 1 kg Plastic powder jar 1kg
10264250 2.5 kg Plastic powder jar 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10637612 Supplier Fisher Chemical Supplier No. P/4080/65
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potassium Carbonate Anhydrous, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Melting Point 891°C
Color White
pH 11.5 to 12
Physical Form Powder/Solid
Quantity 5 kg
IUPAC Name dipotassium carbonate
Formula Weight 138.21g/mol
Packaging Plastic keg
compliance-icons
Product Identifier
  • Potassium carbonate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.